This item is now discontinued. Please browse similar items

Oakton pH Tutor Benchtop Meter with Electrode

Representative image only.
Oakton Item # RK-35619-30
Easy-to-use meter doesn’t sacrifice technology or reliability
  • One of the most economical benchtop meters available
  • Ideal for quick, simple pH measurements
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • Power (VAC)115
  • Power (Hz)60
  • Width (cm)17.78
  • Height (cm)6.0325
  • Width (in)7
  • Height (in)2 3/8
  • DescriptionBenchtop Meter with Electrode
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • One of the most economical benchtop meters available
  • Ideal for quick, simple pH measurements
  • 3 year warranty

More About this Item

Whether you need quick checks in the classroom or laboratory, the pH Tutor provides microprocessor-based technology for fast, accurate readings. The large, dual LCD shows pH measurement and temperature results simultaneously.

Simple push-button keypad allows access to a variety of features, including HOLD, ATC, and electrode diagnostics. The quick reference card, located beneath the meter, slides out for easy reference and keeps procedures handy at all times.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat