Environmental Express® Series 1500 pH Benchtop Meters
Optimized for laboratory measurements
  • Large monochrome LCD
  • Integrated electrode holder (up to 2 electrodes) that can be attached to either side of the meter
  • Simple user interface and single parameter display
  • 1000 Memory data points
  • Automatic Temperature Compensation (ATC) with temperature calibration
  • Adjustable auto shut-off time (1 to 30 minutes)
  • Auto stable, auto hold, and real time measurement modes with reading stability indicators
  • PC (standard USB) / printer (25 pin serial) connection via 2.5 mm diameter phono jack
pH models measure pH and ORP/mV readings. In addition to measurement parameters, each model can be bought as a kit or a meter only (meter only versions include the integrated electrode stand).
$848.67 - $1,060.84CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Benchtop Meter Kit
$1,060.84CAD / Each
COMPARE
pH Benchtop Meter with Electrode Stand
$848.67CAD / Each