METTLER TOLEDO Seven2Go™ Pro S8 Handheld pH/Ion Meter
No more juggling while multitasking
  • Single-handed operation frees a hand for other tasks
  • Robust IP67-rated housing protects meter from harsh conditions
  • Enhanced contrast screen is readable in any light condition—even bright sunlight
  • Compatible with InLab® ISM sensors for quick electrode changes without recalibration
This handheld meter withstands the rigors of busy analytical laboratories, demanding process environments, and unpredictable outdoor applications. The directional keypad with central enter key and intuitive simplified interface allow one hand to set up, calibrate, and measure while the other hand remains free. Waterproof housing resists washdown exposure and inclement weather.
Choose the kit with an electrode and accessories that best fit your application.
$1,628.10 - $2,740.51CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo S8 Handheld pH/Ion Meter Only 3590361 Mfr # 30207874
COMPARE
Handheld pH/Ion Meter Only
$1,628.10CAD / Each
Mettler Toledo S8 Handheld pH/Ion Meter Standard Kit 3590362 Mfr # 30207875
COMPARE
Handheld pH/Ion Meter Standard Kit
$1,981.71CAD / Each
Mettler Toledo S8 Handheld pH/Ion Meter Field Kit 3590363 Mfr # 30207877
COMPARE
Handheld pH/Ion Meter Field Kit
$2,055.38CAD / Each
Mettler Toledo S8 Handheld pH/Ion Meter Biotech Kit 3590364 Mfr # 30207878
COMPARE
Handheld pH/Ion Meter Biotech Kit
$2,217.45CAD / Each
Mettler Toledo S8 Handheld pH/Ion Meter Fluoride Kit 3590365 Mfr # 30207879
COMPARE
Handheld pH/Ion Meter Fluoride Kit
$2,740.51CAD / Each