Horiba B-340 Compact Nitrate Pocket Testers
Flat sensor enables easy measurement with only a few drops of sample!
  • Simply drop the sample directly on the sensor
  • The waterproof IP67-rated design protects from moisture damage for use in wet environments
Only 0.3 mL is needed for a reading. Whether you are determining nitrate concentrations in the field or in the lab, this compact and durable meter provides simple, accurate, and fast measurements. Selectable analysis for NO3 or NO3-N and selectable units of ppm, mg/L, or kg/10a. Automatic calibration function recognizes standards and is user-selectable for either one- or two-point calibrations.
General-purpose nitrate meter includes: 150 and 2000 ppm standards, soft case, and batteries.
Soil nitrate meter includes: 30 and 300 ppm standards, 1-mL droppers, cleaning bottle, extraction bottle, spoons, filter paper, tweezers, hard carrying case, and batteries.
Crop nitrate meter includes: 300 and 5000 ppm standards, 1-mL droppers, cleaning bottle, sample press, sample cups, hard carrying case, and batteries.
$1,234.55 - $2,164.34CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Horiba NO3-11 Compact Nitrate Pocket Tester, general purpose 0576005 Mfr # 3200689162
COMPARE
Compact Nitrate Pocket Tester, general purpose
$1,234.55CAD / Each
Horiba NO3-11C Compact Nitrate Pocket Tester, for crops 0576010 Mfr # 3200689163
COMPARE
Compact Nitrate Pocket Tester, for crops
$1,905.98CAD / Each
Horiba NO3-11S Compact Nitrate Pocket Tester, for soil 0576015 Mfr # 3200689164
COMPARE
Compact Nitrate Pocket Tester, for soil
$2,164.34CAD / Each