YSI PRO 10 Pro 10 pH/ORP/temperature portable meter

Representative image only.

YSI PRO 10 Pro 10 pH/ORP/temperature portable meter

YSI | Mfr: 6050010| Item: 5935216

Configurable to laboratory or field applications

$1,109.46CAD / Each

$1,109.46CAD / Each

Buy It Together

Current Item

YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
5935216

$1,109.46CAD / Each
+
YSI Pro30™ Conductivity Meter
5935205

$1,240.59CAD / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220

$346.25CAD / Each
+ +
YSI Pro 1020 pH and Dissolved Oxygen Meter
5935210

$1,720.92CAD / Each
+ =
Total Price 7894.42 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Includes

two C batteries. Order lab probes with cables; or field probes and cables separately.

Key Features

  • Choose from laboratory probes with cables, or separate field probes and field cable lengths
  • Readable in any lightbacklit display with glow-in-the-dark key pad
  • Tough, IP67 rated waterproof housing with military spec connectors
  • Stores up to 50 data sets in memory
  • 3 year warranty

More About this Item

This easy-to-read meter features a graphic display with detailed messages in user-selectable languages. Backlit display and keypad are ideal for low-light situations. Plus, the rugged rubber case protects meter during field use. Meter is IP67 waterproof rated, even without the battery cover.

The easy-to-use Pro10 allows you to measure your choice of pH or ORP with temperature with probes to suit your application. Meter provides automatic temperature compensation. For laboratory applications, order meter and pH or ORP electrode cable combinations with 1- and 4-m cable lengths. For field applications, order meter, separate pH or ORP electrode, and a 4-, 10-, 20-m field cable.

3 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeGraphic LCD with backlight
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01
  • pH Accuracy±0.2
  • Buffer RecognitionAutomatic
  • Min mV (mV)-1500
  • Max mV (mV)1500
  • mV Resolution1 mV
  • mV Accuracy±20
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)5.715
  • Height (cm)8.255
  • Width (in)2 1/4
  • Length (in)8 1/2
  • Height (in)3 1/4
  • DescriptionPro 10 pH/ORP/temperature portable meter
  • Warranty3 year
Reviews
Q&A