WTW ProfiLine pH 3310 Handheld pH Meters
A rugged new line of electrochemistry meters with a range of features, benefits, and price points to meet every application and budget!
  • Advanced 3310 models feature an innovative waterproof USB connection for ultra-flexible data output
  • Broad pH buffer recognition to meet most calibration needs
  • Rugged, waterproof housing is great for the lab or the field
Durable meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.
pH 3310 meters add extended data memory for logging up to 5000 GLP-compliant data sets. Time-controlled datalogging function offers adjustable data collection intervals—ideal for long-term field monitoring. A special “sleep” mode saves power and extends battery life when performing long-term monitoring. Nonvolatile-memory saves all stored data even if battery power is lost. Data transfer to a PC is made simple via the unique waterproof mini-USB interface.
Meters include: four AA batteries.
Meter kits add: specified probe, calibration solutions, and hard carrrying case.
Meters with drinking water kit further add: Sentix® 81 probe, stand, beaker, measuring vessel, and CD-ROM.
$1,482.23 - $1,863.84CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max pH (pH) Min pH (pH) Description
Availability Pricing
WTW pH 3310 ProfiLine Handheld Meter Only 5889020 Mfr # 2AA310
COMPARE
20 -2 ProfiLine Handheld Meter Only
$1,482.23CAD / Each
WTW pH 3310 ProfiLine meter kit with SenTix 41 pH electrode 5889022 Mfr # 2AA312
COMPARE
20 -2 ProfiLine meter kit with SenTix 41 pH electrode
$1,863.84CAD / Each