Hach HQ440d Benchtop pH and pH/ISE Meter Kits
IntelliCAL™ smart probes have ''plug-and-play'' technology for maximum flexibility
  • IntelliCAL™ probes are precalibrated for maximum accuracy
  • Super-bright LCD and large font for easy-to-read results
  • Option to select from 13 languages in setup mode
  • Store up to 500 data sets for GLP-compliant reporting
Versatile HQd meters offer a large graphic display with backlight, and the intuitive user-interface provides guided calibration, setup, and measurement routines to simplify meter operation. These convenient kits include the dual-input multiparameter meter and probe to measure pH/mV, ISE, and ORP with temperature. Then simply purchase additional IntelliCAL smart probes for other parameters such as conductivity and LDO.
Meter can automatically or manually log up to 500 measurements with probe serial number, current calibration data, user ID, sample ID, time, and date. Use the USB port to download data to a PC or flash drive.
The HQd series uses precalibrated Hach IntelliCAL smart probes that store their own calibration data and identification. The meter recognizes each probe, so you can easily switch probes, and the meter is automatically set up to run the next parameter without recalibrating.
$5,880.30 - $7,166.57CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach pH HQ440D Benchtop Meter Package with PHC281 Ultra Electrode 5900422 Mfr # 8508600
COMPARE
HQ440D Benchtop Meter Package with PHC281 Ultra Electrode
$5,880.30CAD / Each
Hach pH ammonia kit HQ440d Digital Ammonia Bench Meter Kit 5900428 Mfr # 8507600
COMPARE
HQ440d Digital Ammonia Bench Meter Kit
$6,435.77CAD / Each
Hach 8508000 HQ440d Digital Fluoride Bench Meter Kit 5900432 Mfr # 8508000
COMPARE
HQ440d Digital Fluoride Bench Meter Kit
$7,166.57CAD / Each
Hach pH nitrate kit HQ440d Digital Nitrate Bench Meter Kit 5900436 Mfr # 8508100
COMPARE
HQ440d Digital Nitrate Bench Meter Kit
$7,112.05CAD / Each