Extech PH220-C Palm pH Meter with Cabled Electrode

Representative image only.

Extech PH220-C Palm pH Meter with Cabled Electrode

Extech | Mfr: PH220-C| Item: 5302643

Big Digit LCD display in rugged housing with choice of stick or cabled pH Electrode

$322.67CAD / Each

for an additional
$153.97CAD / Each
for an additional
$276.26CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$322.67CAD / Each

Buy It Together

Current Item

Extech PH220-C Palm pH Meter with Cabled Electrode
Extech PH220-C Palm pH Meter with Cabled Electrode
5302643

$322.67CAD / Each
+ +
Extech RE305 Exstik Replacement ORP Electrode
3565003

$117.87CAD / Each
=
Total Price 594.51 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

protective rubber holster with magnetic back, pH electrode, pH 4 & 7 buffers, and 9V battery.

Key Features

  • Large LCD displays pH or mV and temperature simultaneously
  • Microprocessor provides for automatic buffer recognition
  • Memory stores 15 readings
  • Automatic temperature compensation via Pt-100 sensor built into electrode
  • Unit is completely waterproof when electrode is connected to the meter
  • Rugged design for handheld or wall mount
  • Magnetic back panel offers optimum flexibility and convenience
  • Low battery indicator and auto power off
  • 1 year warranty

More About this Item

Specifically designed for fast response measurement.

1 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01
  • pH Accuracy±0.01 pH
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution1 mV
  • mV Accuracy±2 mV
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery1 9V (included)
  • Width (cm)7.897622
  • Height (cm)11.176
  • Width (in)3 7/64
  • Length (in)1 1/2
  • Height (in)4 2/5
  • DescriptionPalm pH Meter with Cabled Electrode
  • Warranty1 year
Reviews
Q&A