Extech FL700 Waterproof Pocket Fluoride Tester

Representative image only.

Extech FL700 Waterproof Pocket Fluoride Tester

Extech | Mfr: FL700| Item: 9935930

Direct reading of total fluoride

$381.61CAD / Each

$381.61CAD / Each

Buy It Together

Current Item

Extech FL700 Waterproof Pocket Fluoride Tester
Extech FL700 Waterproof Pocket Fluoride Tester
9935930

$381.61CAD / Each
+
Extech CL203 Reagent Tablets (10 Pk - 100 Tests)
9959817

$41.25CAD / Each
+ =
Total Price 477.38 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

electrode, protective cap, sample cup with reagent and cap, four 3V CR2032 batteries, and 48" neck strap

Key Features

  • Automatic temperature compensation (±10°C of calibration temperature)
  • Automatic calibration
  • Internal datalogger for storing up to 25 readings
  • LCD displays ppm and temperature with bargraph simultaneously
  • Compliant with EPA Method 340.2 (Potentiometric Ion Selective Electrode)
  • 1 year warranty

More About this Item

This portable, IP57-rated meter is designed for quick and accurate measurement of fluoride ions in drinking water and other aqueous samples. The sensing electrode, measuring electronics, and display are all contained in one easy-to-use package. Meter features auto power-off after 10 minutes and a low-battery indicator.

1 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeLED
  • Min ISE0.10 to 9.99 ppm
  • ISE Resolution0.1 ppm; 0.1 ppt
  • ISE Accuracy±3% or 0.1 ppm
  • WaterproofYes
  • Battery4 3V CR2032 (included)
  • Width (cm)3.65125
  • Height (cm)4.1275
  • Width (in)1 7/16
  • Height (in)1 5/8
  • DescriptionWaterproof Pocket Fluoride Tester
  • Warranty1 year
Reviews
Q&A