Oil Mist Filters
Captures oil residue from exhaust of oil sealed vacuum pumps
  • Seamless drawn housing
  • Prevents oil blow back with auto sealing
  • Easy thumb screw access for maintenance
  • Enclosed housing allows for clean environment
Oil mist filters eliminate oil residue from collecting in your work area. Oil run-off from the filter returns to the pump. Filters contain polyester-wrapped fiberglass elements that coalesce 99.97% of emitted oil. Match the free-air capacity of your pump and outlet port size to the filter. Construction is nickel-chrome plated steel.
$233.53 - $730.43CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Free Air Capacity (CFM) Port Size
Availability Pricing
Rotary Vane Vacuum Pump Oil Mist Filter, 4.5 cfm, 1/2"NPT(M) RK-79775-00 Mfr # EF-FG5-050
COMPARE
4.5 0.5\" NPT(M)
$233.53CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 4.5 cfm, 3/4"NPT(M) RK-79775-04 Mfr # EF-FG5-075
COMPARE
4.5 0.75\" NPT(M)
$275.89CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 4.5 cfm, NW 25 RK-79775-06 Mfr # EF-FG5-NW2516
COMPARE
4.5 NW 25 Flange
$268.53CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 4.5 cfm, NW 16 RK-79775-08 Mfr # EF-FG5-NW16
COMPARE
4.5 NW 16 Flange
$262.63CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 7.0 cfm, 3/4" BSP(M) RK-79775-10 Mfr # EF-FG7-078
COMPARE
7 0.75\" BSP(M)
$262.63CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 7.0 cfm, 1/2"NPT(M) RK-79775-12 Mfr # EF-FG7-050
COMPARE
7 0.5\" NPT(M)
$262.63CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 7.0 cfm, 3/4"NPT(M) RK-79775-14 Mfr # EF-FG7-075
COMPARE
7 0.75\" NPT(M)
$292.47CAD / Each
Rotary Vane Vacuum Pump Oil Mist Filter, 24 cfm, NW 25 RK-79775-20 Mfr # EF-FG10-KF25
COMPARE
24 NW 25 flange
$416.97CAD / Each
Oil mist filter with drain back feature, 16 cfm, NW 25 flange RK-79775-30 Mfr # EFDB-FG9-KF25
COMPARE
16 NW 25 flange
$650.50CAD / Each
Oil mist filterwith drain back feature, 16 cfm, 1"-20 fine(M) RK-79775-35 Mfr # EFDB-FG9-103
COMPARE
16 1\" UNF(M)
$730.43CAD / Each
Showing 1- 10 of 10