Traceable® Certificate, bottle probe with cable, and batteries.
Key Features
Connect to your data via smartphone or tablet—any Bluetooth-enabled device—using a downloadable Bluetooth App
View and download data with free TraceableGO™ App—data can be exported from the TraceableGO™ app and loaded to TraceableLIVE® cloud service*
Data loggers store up to 90 alarm events
Readings and setup parameters easily viewable on large 2" by 1" LCD
Temperature-buffered probe is ideal for monitoring vaccines or other important samples in transit
Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
1 year warranty
More About this Item
Track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals, and food items during transport to any location—Bluetooth® loggers monitor shipments while in transit. Compact, durable data loggers fit almost anywhere—even inside a small cooler.
Configure the data loggers from your device using the TraceableGO™ App to start; stop; change memory mode, unit preference, alarm setting, logging interval; and enable/disable alarm. When all the parameters are set, simply press START on the logger to begin monitoring. Once the shipment is delivered, double press START/STOP on the logger to enable Bluetooth® and export the data to your device. You will get an immediate response to see if your contents have been exposed to conditions outside the set parameters.
Measurements are updated every 5 seconds with user-defined reading intervals from 1 minute to 12 hours in 1-minute steps. Logging duration is 227 days (7.5 months) on a 5-minute logging interval. Data loggers store up to 90 alarm events. Data logger displays current reading, Min/Max, and run time or alarm time.
You have the option to subscribe to cloud-based TraceableLIVE® service to upload data to cloud—ideal for tracking data from multiple loggers or to comply with regulatory agency requirements. The low-cost TraceableLIVE® subscription provides unlimited trip data to be saved on the cloud as well as the ability to send a report to an email.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Related Article:How to Safely Transport Vaccines, how to meet the critical challenge of ensuring a temperature-controlled supply chain from manufacturer to end-user.
Guaranteed to have at least one year of calibration remaining on certificate.
*TraceableLIVE® subscription required to upload data from the TraceableGO™ app.
For sale only in Canada, Europe, and USA.
Specifications
Number of Channels1
Width (InchShort)3 1/2
Input TypeTemperature; bottle probe
Max Temperature (° F)158
Height (InchShort)2
Min Temperature (° F)-58
Max Temperature (° C)70
Min Temperature (° C)-50
Temperature Resolution (° F)0.1°
Temperature Accuracy (° F)±0.5°F
Temperature Resolution (° C)0.1°
Temperature Accuracy (° C)±0.3°C
Sampling Rate1 minute to 12 hours
Memory Size64K (65,536) data points, 7.5 months on 5-minute interval
InterfaceBluetooth
Battery Type2 AAA (included)
Display TypeMultiline LCD
DescriptionTemperature Bluetooth® Enabled Data Logger Compatible with TraceableGO™ App and TraceableLIVE® Cloud Service; Bottle Probe