Traceable Temperature/Humidity Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 1 Dongle
Representative image only.
Traceable Temperature/Humidity Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 1 Dongle
Traceable | Mfr: 6520TR| Item: 1800029
Monitor environmental temperature and humidity from your mobile devices
$625.45CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$625.45CAD / Each
Product Description
Includes
Traceable® Certificate, a dongle with temperature/humidity sensor, built-in stand, and batteries.
Key Features
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*
  • View current humidity and temperature conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
  • One dongle
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
TraceableLIVE Wireless Technology allows multiple users to stay connected and monitor critical environments 24/7 wherever you go.
Remotely monitoring the temperature and humidity of laboratories, warehouses, and more has never been easier! Continuous data transmission using Wi-Fi provides a simple and reliable way to wirelessly monitor the environment.
Once setup is complete you can securely monitor parameters 24/7, control your alarms remotely, view your data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for out-of-range conditions, connectivity interruptions, low-battery condition or if your device is due for calibration.
View current and Min/Max readings on the large tripe-line LCD. A convenient built-in bench stand makes it easy to read the temperature and humidity measurements. Select from model with a dongle or model with external probe and cable.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
*Requires a TraceableLIVE subscription.
For sale only in Australia, Brazil, Canada, China, Europe, India, Japan, Mexico, New Zealand, Singapore, South Korea, and USA. Devices do not support 5 GHz Wi-Fi connections and only connect to 2.4 GHz frequency.
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Cloud ConnectedYes
  • Number of Channels1
  • Input TypeDongle with Temp/Humidity Sensor
  • Max Temperature (° F)131
  • Min Temperature (° F)32
  • Max Temperature (° C)55
  • Min Temperature (° C)0
  • Temperature Resolution (° F)0.01
  • Temperature Accuracy (° F)±0.72
  • Temperature Resolution (° C)0.01
  • Temperature Accuracy (° C)±0.4
  • Max Humidity RH (RH)100
  • Min Humidity RH (RH)0
  • Humidity Resolution (%)0.1%
  • Humidity Accuracy±3% (20 to 80% RH), ±5% rest of range
  • Sampling Rate12 seconds for data logging; 15 minutes default Wi-Fi transmission
  • Memory Size672 records (7 days worth if set to 15 minute interval)
  • InterfaceWi-Fi
  • System RequirementsMeter and TraceableLIVE subscription
  • Alarm100 (stored max)
  • ConnectionsUSB
  • Battery Type4 AAA (included)
  • Display TypeDual-line LCD
  • DescriptionTemperature/Humidity Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 1 Dongle