Traceable Extreme-Accuracy Digital Thermometer, 0°C; 1 Stainless Steel Probe

Representative image only.

Traceable Extreme-Accuracy Digital Thermometer, 0°C; 1 Stainless Steel Probe

Traceable | Mfr: 4241TR| Item: 9000023

Perfect for monitoring critical samples, quality control checks, as well as routine measurements

$237.58CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$237.58CAD / Each

Buy It Together

Current Item

Traceable Extreme-Accuracy Digital Thermometer, 0°C; 1 Stainless Steel Probe
Traceable Extreme-Accuracy Digital Thermometer, 0°C; 1 Stainless Steel Probe
9000023

$237.58CAD / Each
+ + + +
Traceable High-Precision RTD Thermometer
3780392

$691.02CAD / Each
+ + + + + =
Total Price 4332.51 ADD 10 ITEMS TO CART
Product Description

Includes

Traceable® Certificate, stainless steel probe, stand, hook-and-loop fastener, magnetic strips, wall mount, and batteries.

Key Features

  • Accuracy of ±0.05°C within ±2°C of tested points
  • Resolution is 0.01°
  • Displays date and time of min/max temperature
  • High and low settable alarms
  • Unit conforms to ITS-90, International Temperature Standard
  • °C/°F selectable
  • Takes routine measurements at 0.00°C
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

Use extreme-accuracy thermometers for measuring temperatures in water baths, cuvettes, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater. These digital thermometers provide the exact readings required for critical requirements, quality control checks, and routine measurements. Units read 0.00, 25.00, 37.00, or a combination of all three with a resolution of 0.01° and an accuracy ±0.05°C within ±2°C of standard.

View minimum/maximum and current temperature on the triple-line LCD. MIN/MAX monitors readings overnight, on weekends, or for any time period and displays the exact time and date when the MIN/MAX temperature occurred. Alarm provides two visual (LEDs) and two audio alerts when temperature rises above or falls below high and low set points. Unit displays the exact time and date when dual thermometer alarms are triggered. Alarms are programmable in 0.1° increments. Visual and audible alarms signal continuously (for days) even if temperature returns to nonalarm range. First thermometer ever designed to permit user one-button calibration.

Stainless-steel probe supplied with units shows the slightest temperature change. Use probe with liquids, air/gas, and semisolids. Penetration tip probe measures 1/8” dia x 6-1/4”L with overall length of 9”.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)158
  • Min Temperature (° C)-50
  • Max Temperature (° C)70
  • Accuracy±0.05°C when within ±2°C of 0°C; ±1°C otherwise
  • Resolution0.01°
  • AlarmAudible and visual
  • Min/Max FunctionYes
  • Low Battery IndicatorYes
  • Height (in)4 1/4
  • Width (in)2 3/4
  • Depth (in)3/4
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.125
  • Probe Length (in)6 1/4
  • Cable Length (in)60
  • Battery2 AA (included)
  • Probe TypeStainless Steel
  • TestPoints0.00°C
  • DescriptionExtreme-Accuracy Digital Thermometer, 0°C; 1 Stainless Steel Probe
  • Warranty1 year
Reviews
Q&A