Traceable High-Precision RTD Thermometer

Representative image only.

Traceable High-Precision RTD Thermometer

Traceable | Mfr: 4132TR| Item: 3780392

State-of-the-art aerospace technology delivers high platinum RTD performance

$691.02CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$691.02CAD / Each

Buy It Together

Current Item

Traceable High-Precision RTD Thermometer
Traceable High-Precision RTD Thermometer
3780392

$691.02CAD / Each
+ + + +
Waring 019448 Accessory Lid F/04248-30 -60 -90
0424832

$14.88CAD / Each
+ + =
Total Price 2130.59 ADD 7 ITEMS TO CART
Product Description

Includes

Traceable® Certificate, probe, probe holder, hard carrying case, and batteries.

Key Features

  • Enhanced precision with platinum probe
  • Green/Red dual color backlight indicates temperature hazards
  • IP67 waterproof housing—perfect for rugged applications
  • Increase versatility with timer and alarm functions
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

Developed specifically for lab and plant use, the high-performance RTD thermometer has a wide temperature range of -100 to 300°C (-148 to 572ºF) with 0.01° resolution. State-of-the-art, fourth-generation aerospace engineering incorporates the most modern and sophisticated electronics in temperature measurement to achieve utmost in precision, stability, and dependability.

Ensure fast and accurate temperature measurements with real-time response RTD probe; data updates in 1 second time interval. The meter features IP67-rated housing, green/red light HACCP indicator, countdown timer, Min/Max, Hold function to freeze current reading, and high temperature alarm, making it perfect for measuring temperatures indoors, outdoors, and washdown areas.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-148
  • Max Temperature (° F)572
  • Min Temperature (° C)-100
  • Max Temperature (° C)300
  • Accuracy± (0.1% + 0.2°C) below 200°C, ± (0.15% + 0.5°C) otherwise
  • Resolution0.01°
  • AlarmYes
  • IP RatingIP67
  • Height (in)6 5/16
  • Width (in)2 1/16
  • Depth (in)7/8
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.1
  • Probe Length (in)5 29/32
  • Cable Length (in)43
  • Battery4 AAA (included)
  • Probe TypeStainless Steel
  • DescriptionHigh-Precision RTD Thermometer
  • Warranty1 Year
Reviews
Q&A