Traceable® High-Accuracy Digital RTD Thermometers with Calibration
Monitor minimum and maximum temperatures— all at a single glance!
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Temperature-buffered platinum RTD probe
  • RTD probe insures linear response and high accuracy across the entire range
  • Triple-display—shows high, low, and current temperature simultaneously
Easily monitor temperature in liquids, baths, air/gas, incubators, refrigerators, or freezers. Keep track of temperature conditions overnight or for any time period—minimum and maximum temperatures are stored until the memory is cleared. The 4-wire platinum probe insures linear response and high accuracy across the entire range. The built-in alarm provides an alert when temperature rises above or falls below a user defined set point, and is programmable in 1° increments. A visual and audible signal continues even if the temperature returns to non-alarm range until reset by the user. Probe is attached to a 10-ft (3 m) micro-cable that permits a doors to close on it; probe and cable are both submersible. Mount meter with included hook-and-loop strips, wall-mount slots, or use flip-open stand on benchtop.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$241.64 - $270.37CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Description
Availability Pricing
COMPARE
158 Calibrated High-Accuracy RTD Refrigerator/Freezer Digital Thermometer; 1 Bottle Probe
$241.64CAD / Each
COMPARE
158 Calibrated High-Accuracy RTD General Purpose Digital Thermometer; 1 Bullet Probe
$241.64CAD / Each
COMPARE
392 Calibrated High-Accuracy RTD General Purpose Digital Thermometer; 1 Stainless Steel Probe
$270.37CAD / Each
COMPARE
158 Calibrated High-Accuracy RTD General Purpose Digital Thermometer; 1 Bullet Probe
$265.95CAD / Each