Digi-Sense Refrigerator Verification Thermometer; -5 to 15C, PFA Coated
Representative image only.
Digi-Sense Refrigerator Verification Thermometer; -5 to 15C, PFA Coated
Digi-Sense | Item: 9030067
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
$117.58CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$117.58CAD / Each
Buy It Together
Current Item
Digi-Sense Refrigerator Verification Thermometer; -5 to 15C, PFA Coated
Digi-Sense Refrigerator Verification Thermometer; -5 to 15C, PFA Coated
9030067
$117.58CAD / Each
+ + =
Total Price 736.40 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • NIST- traceable statement of accuracy included
  • 5 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. These thermometers are supplied in a media-filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.
5 Year Warranty
Specifications
  • Length (")4 29/32
  • Min Temperature (° C)-5
  • Max Temperature (° C)15
  • Accuracy±0.75°C
  • Division (° C)0.5
  • Thermometer FillBlue Spirit
  • Bottle FillEthylene Glycol
  • Overall Length (InchShort)4.9
  • Overall Length (mm)125
  • Stem MaterialPFA-Coated White-Back Glass
  • DescriptionRefrigerator Verification Thermometer; -5 to 15C, PFA Coated
  • Warranty5 years