Traceable Scientific Thermistor Thermometer with USB Probe

Representative image only.

Traceable Scientific Thermistor Thermometer with USB Probe

Traceable | Mfr: 4000TR| Item: 9008009

A highly accurate thermometer with up to 0.0001° resolution!

$947.39CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$947.39CAD / Each

Buy It Together

Current Item

Product Description

Includes

NIST-traceable calibration certificate with data, general-purpose penetration probe, plastic carrying case, and one 9 V battery.

Traceable® Certificate, 316 stainless steel probe, plastic carrying case, and battery.

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Universal 316 stainless steel probe measure liquids, gases, and semisolids
  • Press the HOLD key to freeze data on the display
  • HISTORY key recalls minimum and maximum readings over the past 24 hours
  • °C/°F selectable
  • Conforms to ITS-90 International Temperature Standard
  • 1 year warranty

More About this Item

Easily measure temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater with this high-precision thermistor thermometer. Expanded temperature range covers –50.000 to 150.000°C (–58.000 to 302.000°F) with resolution of 0.0001° between –9.9999 to 99.9999°. A push of a single button changes the display to show one, two, three, or four significant digits after the decimal point. The fast-response general-purpose probe (included) detects even the slightest temperature change instantly. Readings are updated three times a second.

Store up to 50 temperature measurements with the min/max temperature measurement for the past 24-hours period. The internal memory can be downloaded into a computer using optional software 90080-21 for analysis. The meter features a splash-proof membrane keypad to provide years of reliable service even in severe environments. A high-contrast 1/2"-high LCD makes the thermometer easy to read in all types of light.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyNIST Traceable Calibration
Specifications
  • Meter Measurement TypeThermistor
  • Width (InchShort)3 1/2
  • Number Of Probe Channels1
  • Height (InchShort)1 1/4
  • Types Of Probes AcceptedThermistor
  • Min Temperature (° F)-58
  • Max Temperature (° F)302
  • Min Temperature (° C)-50
  • Max Temperature (° C)150
  • Accuracy±0.05°C between 0 to 100°C
  • Resolutionfrom -50 to 150°C (-50 to 302°F): 0.001°
    from -9.9999 to 99.9999°: 0.0001°
  • OutputUSB
  • Data Logging50 readings
  • Min/Max FunctionYes
  • Low Battery IndicatorYes
  • Display Type1/2" high-contrast LCD, 6-digits
  • Length (in)5 1/2
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.13
  • Probe Length (in)9
  • Cable Length (in)66
  • Stem Length (in)6 19/64
  • Battery Life1 year
  • Battery1 9V (included)
  • Probe TypeThermistor
  • DescriptionCalibrated Scientific Thermistor Thermometer; USB Probe
  • Warranty1 Year
Reviews
Q&A