InnoCal NIST-Traceable Calibration; Timer/Stopwatch
Representative image only.
InnoCal NIST-Traceable Calibration; Timer/Stopwatch
InnoCal | Item: 1706000
Quick and accurate timer calibrations accredited to ISO/IEC 17025 standards
$116.69CAD / Each
$116.69CAD / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; Timer/Stopwatch
InnoCal NIST-Traceable Calibration; Timer/Stopwatch
1706000
$116.69CAD / Each
+
Traceable Three Channel Alarm Timer
9441110
$106.82CAD / Each
+
Traceable Three-Button Stopwatch
9446006
$22.25CAD / Each
+
Traceable Big-Digit Stopwatch
9446028
$63.06CAD / Each
+ +
Traceable 60-Memory Digital Stopwatch
9876602
$81.63CAD / Each
+
Traceable Digital Stopwatch
9876603
$140.27CAD / Each
+ +
Traceable 99min/59s Timer
9876678
$34.92CAD / Each
=
Total Price 737.74 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specifications, EPA, USDA, ISO and GLP/GMPs standards
More About this Item
Calibrations on timers and stopwatches are performed using a precision timometer. Measurements can be taken from 60 seconds to 720 hours.
Calibrations on timers and stopwatches are performed using a precision timometer. Measurements can be taken from 60 seconds to 720 hours.
NIST Traceable Calibration
Specifications
  • Calibration TypeMechanical
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1 Point
  • Calibration Test PointsDeviation of Time-Base
  • DescriptionNIST-Traceable Calibration; Timer/Stopwatch