Solar Light Radiometer Sensors

Replacement sensors for your Solar Light radiometer.
$1,834.37 - $3,160.42CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Intensity Range Description
Availability Pricing
Solar Light PMA2101 UV-B sensor for single-input datalogging radiometer 9765050 Mfr # PMA2101
COMPARE
1160 uW/cm2 UV-B sensor for single-input datalogging radiometer
$2,460.56CAD / Each
Solar Light PMA2110 UV-A sensor for single- or dual-input datalogging radiometer 9765052 Mfr # PMA2110
COMPARE
200 mW/cm2, 2000 W/m2 UV-A sensor for single- or dual-input datalogging radiometer
$2,416.36CAD / Each
Solar Light PMA2130 Visible Phototopic Sensor 1-13/16"H x 1-9/16" dia 9765054 Mfr # PMA2130
COMPARE
220 W/m2 Visible Phototopic Sensor 1-13/16"H x 1-9/16" dia
$3,160.42CAD / Each
Solar Light PMA2120 UV Safety Sensor for Measuring Efeective Actinic UV Irradiance 9765059 Mfr # PMA2120
COMPARE
1000 uW/cm2; 10,000 mW/cm2 UV Safety Sensor for Measuring Efeective Actinic UV Irradiance
$2,460.56CAD / Each
Solar Light PMA2121 Visible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia 9765060 Mfr # PMA2121
COMPARE
2000 uW/cm2; 20,000 mW/cm2 Visible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia
$1,834.37CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat