Solar Light PMA2121 Visible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia

Representative image only.

Solar Light PMA2121 Visible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia

Solar Light | Mfr: PMA2121| Item: 9765060

$1,834.37CAD / Each

$1,834.37CAD / Each

Buy It Together

Current Item

Solar Light PMA2121 Visible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia
Solar Light PMA2121 Visible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia
9765060

$1,834.37CAD / Each
+ =
Total Price 10682.08 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Replacement sensors for your Solar Light radiometer.

Measures the blue light (near UV) radiation hazard to eyes. Can be used by employers and safety officers to protect against the effects of excessive daily blue light exposure.
Weatherproof detector housing available.

NIST Traceable Calibration1 Year Warranty
Specifications
  • Resolution0.01 µW/cm2, 0.1 mW/m2
  • FunctionMeasures blue light (near UV) radiation hazardous to eyes
  • Intensity Range2000 uW/cm2; 20,000 mW/cm2
  • Energy Range2000 µW/cm2 (20,000mW/m2)
  • Depth (in)1 9/16
  • Height (in)1 13/16
  • Height (cm)4.6
  • Depth (cm)3.96
  • DescriptionVisible Blue Light Safety Sensor 1-13/16"H x 1-9/16" dia
  • Warranty1 year
Reviews
Q&A