Solar Light PMA2130 Visible Phototopic Sensor 1-13/16"H x 1-9/16" dia

Representative image only.

Solar Light PMA2130 Visible Phototopic Sensor 1-13/16"H x 1-9/16" dia

Solar Light | Mfr: PMA2130| Item: 9765054

$3,160.42CAD / Each

$3,160.42CAD / Each

Buy It Together

Current Item

Solar Light PMA2130 Visible Phototopic Sensor 1-13/16"H x 1-9/16" dia
Solar Light PMA2130 Visible Phototopic Sensor 1-13/16"H x 1-9/16" dia
9765054

$3,160.42CAD / Each
+ =
Total Price 12008.13 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Replacement sensors for your Solar Light radiometer.

Measures light intensity below 0.01 lux and can also be used as a safety detector for hazardous intensities of light.
Weatherproof detector housing available.

NIST Traceable Calibration1 Year Warranty
Specifications
  • Max Measurement (Lux)150000
  • Resolution1 Lux, 0.001 W/m2
  • FunctionMeasures light intensity below 0.01 Lux, can be used as a safety detector for hazardous intensities of light
  • Intensity Range220 W/m2
  • Energy Range150,000 lux (220 W/m2)
  • Height (in)1 13/16
  • Depth (in)1 9/16
  • Height (cm)4.6
  • Depth (cm)3.96
  • DescriptionVisible Phototopic Sensor 1-13/16"H x 1-9/16" dia
  • Warranty1 year
Reviews
Q&A