Solar Light PMA2120 UV Safety Sensor for Measuring Efeective Actinic UV Irradiance

Representative image only.

Solar Light PMA2120 UV Safety Sensor for Measuring Efeective Actinic UV Irradiance

Solar Light | Mfr: PMA2120| Item: 9765059

$2,460.56CAD / Each

$2,460.56CAD / Each

Buy It Together

Current Item

Solar Light PMA2120 UV Safety Sensor for Measuring Efeective Actinic UV Irradiance
Solar Light PMA2120 UV Safety Sensor for Measuring Efeective Actinic UV Irradiance
9765059

$2,460.56CAD / Each
+ =
Total Price 11308.27 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Replacement sensors for your Solar Light radiometer.

Measures the amount of UV radiation that it is being exposed to and indicates the level at which is unsafe by the standards of the American Conference of Governmental Industrial Hygienists (ACGIH).
Weatherproof detector housing available.

NIST Traceable Calibration1 Year Warranty
Specifications
  • Resolution0.01 µW/cm2, 0.1 mW/m2
  • FunctionMeasures UV radiation and indicates the level which is unsafe by the standards of the American Conference of Governmental Industrial Hygienists (ACGIH).
  • Intensity Range1000 uW/cm2; 10,000 mW/cm2
  • Energy Range1000 µW/cm2, 10,000 mW/m2 (autoranging)
  • Height (in)1 13/16
  • Depth (in)1 9/16
  • Height (cm)4.6
  • Depth (cm)3.96
  • DescriptionUV Safety Sensor for Measuring Efeective Actinic UV Irradiance
  • Warranty1 year
Reviews
Q&A