Regenerative Blowers
  • Quietly recirculate air at up to 420 cfm
  • TEFC motor mounted
  • Use in any orientation to meet your system requirements
  • Perfect for pond or plate bath aeration, wastewater treatment, and trim and dunnage removal
Cord and plug not included.
$1,337.84 - $3,793.98CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Pressure (PSI) Max Vacuum
Availability Pricing
Gast R1102 Regenerative blower, 27 cfm, 115/230 VAC RK-07047-00 Mfr # R1102K-01
COMPARE
27 0.957 28.5\" H2O
$1,337.84CAD / Each
Gast R2103 Regenerative blower, 42 cfm, 115/230 VAC RK-07047-10 Mfr # R2103NX
COMPARE
42 1.264 39\" H2O
$1,351.10CAD / Each
Gast R3105-1 Regenerative blower, 53 cfm, 115/230 VAC RK-07047-20 Mfr # R3105NX
COMPARE
53 1.553 43\" H2O
$1,679.66CAD / Each
Gast R4110-2 Regenerative Blower, Single Phase; 74 cfm at 50 Hz, 92 cfm at 60 Hz RK-07047-30 Mfr # R4110-2
COMPARE
74/92 1.372 34\" H20, 48\"H20
$2,688.94CAD / Each
Gast R5325A-2 Regenerative Blower, Three Phase; 133 cfm at 50 Hz, 160 cfm at 60 Hz RK-07047-45 Mfr # R5325A-2
COMPARE
133/160 1.806 47\" H20, 60\"H20
$2,659.47CAD / Each
Gast R6350A-2 Regenerative Blower, Three Phase; 180 cfm at 50 Hz, 215 cfm at 60 Hz RK-07047-55 Mfr # R6350A-2
COMPARE
180/215 2.817 70\" H20, 88\"H20
$3,793.98CAD / Each