Pyrex® Brand 1060 Berzelius Glass Beakers
Save storage space
  • Beakers are taller instead of wider
  • Easy-read graduations for approximate measuring
  • Easy-pour spout to facilitate sampling
Pyrex® Brand 1060 Berzelius beakers are tall and feature a spout to facilitate easy pouring. Beakers are graduated to indicate their approximate content. Reference: ASTM E-960.
$122.29 - $589.36CAD / Pkg of 12
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 1060-300 Brand 1060 Berzelius Tall Form Beaker, 300 mL, 12/pk RK-34513-03 Mfr # 1060-300
COMPARE
300 25 Glass
$191.54CAD / Pkg of 12
Pyrex 1060-500 Brand 1060 Berzelius Tall Form Beaker, 500 mL, 6/pk RK-34513-05 Mfr # 1060-500
COMPARE
500 50 Glass
$122.29CAD / Pkg of 6
Pyrex 1060-1L Brand 1060 Berzelius Tall Form Beaker, 1000 mL, 6/pk RK-34513-10 Mfr # 1060-1L
COMPARE
1000 50 Glass
$206.27CAD / Pkg of 6
Pyrex 1060-100 Brand 1060 Berzelius Tall Form Beaker, 100 mL, 48/cs RK-34513-21 Mfr # 1060-100
COMPARE
100 10 Glass
$500.95CAD / Case of 48
Pyrex 1060-200 Brand 1060 Berzelius Tall Form Beaker, 200 mL, 48/cs RK-34513-22 Mfr # 1060-200
COMPARE
200 25 Glass
$530.42CAD / Case of 48
Pyrex 1060-300 Brand 1060 Berzelius Tall Form Beaker, 300 mL, 48/cs RK-34513-23 Mfr # 1060-300
COMPARE
300 25 Glass
$589.36CAD / Case of 48
Pyrex 1060-400 Brand 1060 Berzelius Tall Form Beaker, 400 mL, 36/cs RK-34513-24 Mfr # 1060-400
COMPARE
400 25 Glass
$478.85CAD / Case of 36
Pyrex 1060-600 Brand 1060 Berzelius Tall Form Beaker, 600 mL, 24/cs RK-34513-26 Mfr # 1060-600
COMPARE
600 50 Glass
$456.75CAD / Case of 24
Pyrex 1060-1L Brand 1060 Berzelius Tall Form Beaker, 1000 mL, 18/cs RK-34513-30 Mfr # 1060-1L
COMPARE
1000 50 Glass
$456.75CAD / Case of 18
Showing 1- 9 of 9