Pyrex® Brand 1003 Heavy-Duty Glass Beakers
Get the best strength even under harsh conditions
  • Know your solution—extra-large marking spot for easier identification
  • Beakers designed for the harshest conditions
Heavy-duty beakers are specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. The 250-mL to 4000-mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot. Reference: ASTM E-960.
$212.17 - $1,016.64CAD / Pkg of 1
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 1003-150 Brand 1003 Griffin Heavy-Duty Beaker, 150 mL, 12/pk 3451206 Mfr # 1003-150
COMPARE
150 20 Glass
$235.74CAD / Pkg of 12
Pyrex 1003-250 Brand 1003 Griffin Heavy-Duty Beaker, 250 mL, 12/pk 3451207 Mfr # 1003-250
COMPARE
250 25 Glass
$277.00CAD / Pkg of 12
Pyrex 1003-400 Brand 1003 Griffin Heavy-Duty Beaker, 400 mL, 12/pk 3451208 Mfr # 1003-400
COMPARE
400 25 Glass
$350.67CAD / Pkg of 12
Pyrex 1003-600 Brand 1003 Griffin Heavy-Duty Beaker, 600 mL, 6/pk 3451209 Mfr # 1003-600
COMPARE
600 50 Glass
$212.17CAD / Pkg of 6
Pyrex 1003-1L Brand 1003 Griffin Heavy-Duty Beaker, 1000 mL 3451211 Mfr # 1003-1L
COMPARE
1000 50 Glass
$400.76CAD / Pkg of 6
Pyrex 1003-2L Brand 1003 Griffin Heavy-Duty Beaker, 2000 mL, 4/pk 3451213 Mfr # 1003-2L
COMPARE
2000 200 Glass
$521.58CAD / Pkg of 4
Pyrex 1003-4L Brand 1003 Griffin Heavy-Duty Beaker, 4000 mL, 1/pk 3451215 Mfr # 1003-4L
COMPARE
4000 250 Glass
$268.16CAD / Pkg of 1
Pyrex 1003-250 Brand 1003 Griffin Heavy-Duty Beaker, 250 mL, 48/cs 3451227 Mfr # 1003-250
COMPARE
250 25 Glass
$701.33CAD / Case of 48
Pyrex 1003-400 Brand 1003 Griffin Heavy-Duty Beaker, 400 mL, 48/cs 3451228 Mfr # 1003-400
COMPARE
400 25 Glass
$914.98CAD / Case of 48
Pyrex 1003-1L Brand 1003 Griffin Heavy-Duty Beaker, 1000 mL SKU 3451231 3451231 Mfr # 1003-1L
COMPARE
1000 50 Glass
$1,016.64CAD / Case of 24
Pyrex 1003-2L Brand 1003 Griffin Heavy-Duty Beaker, 2000 mL, 8/cs 3451233 Mfr # 1003-2L
COMPARE
2000 200 Glass
$693.97CAD / Case of 8
Pyrex 1003-4L Brand 1003 Griffin Heavy-Duty Beaker, 4000 mL, 4/cs 3451235 Mfr # 1003-4L
COMPARE
4000 250 Glass
$770.58CAD / Case of 4
Showing 1- 12 of 12