Cole-Parmer Sterilized Air Filter; 0.2µm Pore, 50mm Dia, 1/Pk

Representative image only.

Cole-Parmer Sterilized Air Filter; 0.2µm Pore, 50mm Dia, 1/Pk

Cole-Parmer | Item: 1594523

Sterile filter with hydrophobic PTFE membrane for degassing bioreactors

$49.73CAD / Each

$49.73CAD / Each
Product Description

Key Features

  • Stepped hose barb connections accept 1/4" to 3/8" inner diameter tubing

More About this Item

This venting disc filter has a hydrophobic PTFE membrane and PP housing—making it ideal for venting and degassing fermentation tanks and culture vessels, drying or sterilizing gases, and cleaning analytical solvents and reagents. Filter can withstand up to 30 autoclave cycles at 121°C for 20 minutes or 3 cycles at 134°C for 30 minutes.

ISO9001 Certified Supplier
Specifications
  • Membrane MaterialPolytetrafluoroethylene (PTFE)
  • Pore Size (µm)0.2
  • Membrane Diameter (mm)50
  • Housing MaterialPolypropylene (PP)
  • SterileYes
  • Connections1/4" to 3/8" (6 to 9.5 mm) hosebarbs
  • Effective Filtration Area (cm²)16.2
  • Max Pressure (PSI)60
  • Max Pressure (bar)4.1
  • DescriptionSterile Air Vent Filters; 0.2µm Pore, 50mm Dia, 1/Pk
Reviews
Q&A