Cole-Parmer® PTFE Chromatography Syringe Filters
Best suited for analytical and research applications and organic solvent filtration
  • Bidirectional flow with female luer lock on one side and a male luer slip on the other
  • Withstand pressure up to 100 psi (7 bar)
  • Batch test certificate included with each pack
PTFE Syringe Filters stand up to the most aggressive solvent, acid, and base samples and are ideal for filtration of highly aggressive colloidal solutions. These filters can be used under extreme chemical or temperature conditions and are ideal for analytical and research applications where non-aqueous/organic solvent filtration is required. Maximum use temperature is 60°C (140°F).
$123.18 - $228.60CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
13 Polytetrafluoroethylene (PTFE) 0.2
$228.60CAD / Each
COMPARE
25 Polytetrafluoroethylene (PTFE) 0.2
$123.18CAD / Each
COMPARE
25 Polytetrafluoroethylene (PTFE) 0.45
$216.59CAD / Each