Idex® Short Head Male Nuts, 1/4-28 Flat-Bottom Connection for 1/8" OD Tubing
Excellent for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
$2.36 - $55.69CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Flangeless Male Nut, Short Head, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201994 Mfr # P-308
COMPARE
Black Acetal Flangeless Male Nut, Short Head, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
$2.36CAD / Each
Idex Flangeless Male Nut, Short Head, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202004 Mfr # P-318
COMPARE
Natural ETFE Flangeless Male Nut, Short Head, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$3.83CAD / Each
Idex Flangeless Male Nut, Short Head, Black ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202006 Mfr # P-318BLK
COMPARE
Black ETFE Flangeless Male Nut, Short Head, Black ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$5.01CAD / Each
Idex Flangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202009 Mfr # P-318Y
COMPARE
Yellow ETFE Flangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
$8.40CAD / Pkg of 1
Idex Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202023 Mfr # P-335
COMPARE
Natural PEEK Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$5.75CAD / Each
Idex Flangeless Male Nut, Short Head, Blue PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202024 Mfr # P-335B
COMPARE
Blue PEEK Flangeless Male Nut, Short Head, Blue PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$6.04CAD / Each
Idex Flangeless Male Nut, Short Head, Green PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202025 Mfr # P-335G
COMPARE
Green PEEK Flangeless Male Nut, Short Head, Green PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$7.22CAD / Each
Idex Flangeless Male Nut, Short Head, Orange PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202026 Mfr # P-335OR
COMPARE
Orange PEEK Flangeless Male Nut, Short Head, Orange PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$7.51CAD / Each
Idex Flangeless Male Nut, Short Head, Red PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202027 Mfr # P-335R
COMPARE
Red PEEK Flangeless Male Nut, Short Head, Red PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$7.22CAD / Each
Idex Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202028 Mfr # P-335X
COMPARE
Natural PEEK Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
$55.69CAD / Pkg of 10
Idex Flangeless Male Nut, Short Head, Yellow PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202029 Mfr # P-335Y
COMPARE
Yellow PEEK Flangeless Male Nut, Short Head, Yellow PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$7.22CAD / Each
Showing 1- 11 of 11