Yamato Gravity Convection Ovens
Fast drying times — dual exhaust ports move air efficiently
  • Feature calibration functions perfect for validation work
Set temperatures easily and accurately with the push-button digital controller. LED display shows set temperature, actual temperature, and overtemperature set point. Self-diagnostic microprocessor PID monitors temperature and visually and audibly alerts you to sensor or heater malfunction.
$2,921.73 - $5,909.77CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Yamato DX-302 High-Temperature Gravity Convection Oven, 0.9 Cu Ft, 115 VAC 3395900 Mfr # DX-302C
COMPARE
0.9 115
$2,921.73CAD / Each
Yamato DX-402 High-Temperature Gravity Convection Oven, 2.6 Cu Ft, 115 VAC SKU 3395901 3395901 Mfr # DX-402C
COMPARE
2.6 115
Call For Price
Yamato DX-602 High-Temperature Gravity Convection Oven, 5.4 Cu Ft, 115 VAC SKU 3395902 3395902 Mfr # DX-602C
COMPARE
5.4 115
$5,064.04CAD / Each
Yamato DVS-402 Programmable Gravity Convection Oven, 3.5 Cu Ft, 115VAC 3395903 Mfr # DVS-402C
COMPARE
3.5 115
$4,750.21CAD / Each
Yamato DVS-602 Programmable Gravity Convection Oven, 5.6 Cu Ft, 115VAC 3395904 Mfr # DVS-602C
COMPARE
5.6 115
$5,909.77CAD / Each