Swift Optical M17 Series Infinity-Corrected Compound Microscopes
Infinity-corrected optics maintain parfocality even when an auxiliary component is added—perfect for advanced studies and professional applications
  • Ergonomic 30° inclined Siedentopf head provides operator comfort
  • 20 mm eyepieces increase field of view
  • Reversed nosepiece allows clear access to stage
  • Smooth, ball-bearing mounted quad objectives for precise positioning of objectives
  • Shielded iris diaphragm for added protection
  • All metal internal parts offer durability
The M17 series compound microscopes feature infinity-corrected optics that allow one or more auxiliary components (illuminators or polarizer) to be added without altering the alignment between the objective and tube lens. Even with added components infinity-corrected optics produce exactly 1x magnification allowing easy comparison between specimens.
These research level microscopes feature a 30° inclined eyepieces with diopter and interpupilllary adjustment from 54 to 76 mm. A bright 3 W LED illumination system and widefield 20 mm eyepieces provide a sharp image. The 100xR objective is spring-loaded and sealed to protect optics from contact or oil damage. Optically coated Abbe 1.25 N.A. brightfield condenser allows maximum transmission and enhanced contrast. Large 14.2 x 14 mm (5-3/4" x 5-1/2"), low-drive mechanical stage offers precise scanning of your specimen.
$1,663.46 - $3,692.32CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Objectives Description
Availability Pricing
Swift Optical M17B-P Infinity-Corrected Compound Microscope, Binocular, Plan 4x/10x/40xR/100xR RK-48926-72 Mfr # M17B-P
COMPARE
Binocular Infinity-corrected plan objectives 4x, 10x, 40xR, and 100xR Infinity-Corrected Compound Microscope, Binocular, Plan 4x/10x/40xR/100xR
$1,663.46CAD / Each
Swift Optical M17B-MP Infinity-Corrected Compound Microscope, Binocular, Plan 4x/100xR & Phase 10x/40xR RK-48926-73 Mfr # M17B-MP
COMPARE
Binocular Infinity-corrected plan objectives 4x and 100xR; infinity phase objectives 10x and 40xR Infinity-Corrected Compound Microscope, Binocular, Plan 4x/100xR & Phase 10x/40xR
$2,890.79CAD / Each
Swift Optical M17T-P Infinity-Corrected Compound Microscope, Trinocular, Plan 4x/10x/40xR/100xR RK-48926-74 Mfr # M17T-P
COMPARE
Trinocular Infinity-corrected plan objectives 4x, 10x, 40xR, and 100xR Infinity-Corrected Compound Microscope, Trinocular, Plan 4x/10x/40xR/100xR
$2,140.84CAD / Each
Swift Optical M17T-MP Infinity-Corrected Compound Microscope, Trinocular, Plan 4x/100xR & Phase 10x/40xR RK-48926-75 Mfr # M17T-MP
COMPARE
Trinocular Infinity-corrected plan objectives 4x and 100xR; infinity phase objectives 10x and 40xR Infinity-Corrected Compound Microscope, Trinocular, Plan 4x/100xR & Phase 10x/40xR
$3,356.38CAD / Each
Swift Optical M17T-BTI1-P Infinity-Corrected Compound Microscope, Trinocular, Plan Optics, WiFi Camera, 8" Tablet RK-48926-76 Mfr # M17T-BTI1-P
COMPARE
Trinocular Infinity-corrected plan objectives 4x, 10x, 40xR, and 100xR Infinity-Corrected Compound Microscope, Trinocular, Plan Optics, WiFi Camera, 8\" Tablet
$3,245.88CAD / Each
Swift Optical M17T-BTI2-P Infinity-Corrected Compound Microscope, Trinocular, Plan Optics, WiFi Camera, 10" Tablet RK-48926-77 Mfr # M17T-BTI2-P
COMPARE
Trinocular Infinity-corrected plan objectives 4x, 10x, 40xR, and 100xR Infinity-Corrected Compound Microscope, Trinocular, Plan Optics, WiFi Camera, 10\" Tablet
$3,692.32CAD / Each