Abdos™ Polypropylene Beakers with Handle
No more awkward transfers
  • Rectangular handle and spout make pouring easy
  • Highly visible black graduations for approximate volume
  • Autoclavable
The polypropylene (PP) beakers are durable and chemically-resistant making them ideal for general laboratory use. They can withstand temperatures to 250°F (121°C).
$47.30 - $133.93CAD / Pkg of 6
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Abdos Beaker with Handle, PP, 100 mL; 6/pk 6100046 Mfr # P50802
COMPARE
100 10 Polypropylene(PP)
$47.30CAD / Pkg of 6
Abdos Beaker with Handle, PP, 250 mL; 6/pk 6100047 Mfr # P50803
COMPARE
250 25 Polypropylene(PP)
$59.23CAD / Pkg of 6
Abdos Beaker with Handle, PP, 500 mL; 6/pk 6100048 Mfr # P50804
COMPARE
500 50 Polypropylene(PP)
$81.92CAD / Pkg of 6
Abdos Beaker with Handle, PP, 1000 mL; 4/pk 6100049 Mfr # P50805
COMPARE
1000 50 Polypropylene(PP)
$90.61CAD / Pkg of 4
Abdos Beaker with Handle, PP, 2000 mL; 4/pk 6100054 Mfr # P50806
COMPARE
2000 100 Polypropylene(PP)
$133.93CAD / Pkg of 4