A&D Weighing Weigh-Below Device Kits
Expand the uses of your balance with the below-balance weighing hook
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.
$103.14 - $360.98CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
A&D Weighing EJ-07 Under Hook for EJ-3000/4100/6100 Balances 1111504 Mfr # EJ-07
COMPARE
UndeRH/ook for EJ-3000/4100/6100,
$132.61CAD / Each
A&D Weighing EJ-08 Under Hook for EJ-1500/2000 Balances 1111505 Mfr # EJ-08
COMPARE
UndeRH/ook for EJ-1500/2000,
$132.61CAD / Each
A&D Weighing EKW-07I Under Hook for Ek-4100I, -6100I, 6000I, -12Ki, Ew-12Ki, and Ekw-07I Balances 1111508 Mfr # EKW-07I
COMPARE
UndeRH/ook for Ek-4100I / 6100I / 6000I / 12Ki / Ew-12Ki, Ekw-07I
$103.14CAD / Each
A&D Weighing GP-20 Underhook Option (12Kg through 40Kg) 1111538 Mfr # GP-20
COMPARE
UndeRH/ook Gp-12kg Thru 40kg,
$360.98CAD / Each
A&D Weighing GP-21 Underhook Option (60Kg through 102Kg) 1111539 Mfr # GP-21
COMPARE
UndeRH/ook Gp-60kg Thru 102kg,
$360.98CAD / Each
A&D Weighing GP-20 Under Hook for Gp-12kg thru 41kg Balances 1112698 Mfr # GP-20
COMPARE
Balance UndeRH/ook 12kg-41kg
$360.98CAD / Each
A&D Weighing GP-21 Under Hook for Gp-61kg thru 101kg Balances 1112699 Mfr # GP-21
COMPARE
Balance UndeRH/ook 61kg-101kg
$360.98CAD / Each
Showing 1- 7 of 7