A&D Weighing EJ-08 Under Hook for EJ-1500/2000 Balances
Representative image only.
A&D Weighing EJ-08 Under Hook for EJ-1500/2000 Balances
A&D Weighing | Mfr: EJ-08| Item: 1111505
Expand the uses of your balance with the below-balance weighing hook
$132.61CAD / Each
$132.61CAD / Each
Product Description
Key Features
  • Compatible with A&D EJ-1500/2000 Balances
  • 3 month warranty
More About this Item
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.
3 Month WarrantyConformite Europeenne
Specifications
  • DescriptionUndeRH/ook for EJ-1500/2000,