A&D Weighing GP-21 Underhook Option (60Kg through 102Kg)
Representative image only.
A&D Weighing GP-21 Underhook Option (60Kg through 102Kg)
A&D Weighing | Mfr: GP-21| Item: 1111539
Expand the uses of your balance with the below-balance weighing hook
$360.98CAD / Each
$360.98CAD / Each
Product Description
Key Features
  • Compatible with A&D Gp-60kg Thru 102kg Balances
  • 3 month warranty
More About this Item
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.
3 Month WarrantyConformite Europeenne
Specifications
  • DescriptionUndeRH/ook Gp-60kg Thru 102kg,