A&D Weighing EKW-07I Under Hook for Ek-4100I, -6100I, 6000I, -12Ki, Ew-12Ki, and Ekw-07I Balances
Representative image only.
A&D Weighing EKW-07I Under Hook for Ek-4100I, -6100I, 6000I, -12Ki, Ew-12Ki, and Ekw-07I Balances
A&D Weighing | Mfr: EKW-07I| Item: 1111508
Expand the uses of your balance with the below-balance weighing hook
$103.14CAD / Each
$103.14CAD / Each
Product Description
Key Features
  • Compatible with A&D Ek-4100I, -6100I, 6000I, -12Ki, Ew-12Ki, and Ekw-07I Balances
  • 3 month warranty
More About this Item
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.
3 Month WarrantyConformite Europeenne
Specifications
  • DescriptionUndeRH/ook for Ek-4100I / 6100I / 6000I / 12Ki / Ew-12Ki, Ekw-07I