YSI Pro 1020 pH and Dissolved Oxygen Meter

Representative image only.

YSI Pro 1020 pH and Dissolved Oxygen Meter

YSI | Mfr: 6051020| Item: 5935210

Rugged and reliable handheld instrument

$1,720.92CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,720.92CAD / Each

Buy It Together

Current Item

YSI Pro 1020 pH and Dissolved Oxygen Meter
YSI Pro 1020 pH and Dissolved Oxygen Meter
5935210

$1,720.92CAD / Each
+
YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
5935216

$1,109.46CAD / Each
+ + =
Total Price 3475.35 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

two C alkaline batteries. Order probes and cable separately.

Key Features

  • IP67-rated waterproof housing with military-spec connectors
  • Large backlit display and glow-in-the dark keypad for any lighting condition
  • 50-data set memory; no need to write down
  • Combine meter with pH and DO probes, and cable lengths to meet your needs
  • 3 year warranty

More About this Item

The Pro1020 line allows you to measure temperature, pH or ORP, and DO with automatic barometric compensation. Easy-to-use meters feature graphic display with detailed messages to guide the user. Backlit display and keypad are ideal for low-light situations. Rugged rubber case protects meters during use. Meters are IP67 waterproof rated,with military-spec connectors for the hashest field conditions.
Meter provides automatic temperature compensation when pH is measured.

IP 67ISO9001:2000 Certified Supplier3 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeFour-line backlit LCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01
  • pH Accuracy±0.2
  • Buffer RecognitionNIST and USA
  • Min mV (mV)-1500
  • Max mV (mV)1500
  • mV Resolution1 mV
  • mV Accuracy±20
  • Min Dissolved Oxygen0 to 500%; 0 to 50 ppm
  • Dissolved Oxygen Resolution0.01 ppm; 0.1% saturation
  • Dissolved Oxygen Accuracy±2% readinr or ±0.2 whichever is greater
  • Min Temperature (° C)-5
  • Max Temperature (° C)55
  • Min Temperature (° F)23
  • Max Temperature (° F)131
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery2 C (included)
  • Battery life (hr)1000
  • Width (cm)8.255
  • Height (cm)21.59
  • Width (in)3 1/4
  • Length (in)2 1/4
  • Height (in)8 1/2
  • DescriptionpH and Dissolved Oxygen Meter
  • Warranty3 year
Reviews
Q&A