YSI ProSolo Optical Dissolved Oxygen Meter
Get lab-quality results at the source
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Robust IP67-rated housing and connectors withstands harsh field conditions Smart probes store calibration information for up to one year saving you time and money Store more than 100,000 datasets in onboard memory for later download to PC or tablet Order kit or handheld meter and probes separately
Perfectly suited for the natural world, these optical dissolved oxygen meters combine durable water-proof construction with a user-friendly interface and the flexibility of using existing probes or upgrading to the next generation. Because there is no need to stir to get an accurate reading, the probe technology is ideal for low-agitation applications such as deep-water profiling. Meter features an expansive memory with storage space for over 100,000 datasets, reducing dependence on proximity to a computer—included flash drive lets you stay out in the field even longer. KorDSS data acquisition and management software simplifies analysis.
Interchangeable digital "smart" probes store calibration data for a year, saving you time and calibration solution costs—no need to recalibrate if connected to different meters. Convenient cable management kits are included with probes featuring cable lengths from 13-ft (4-m) to 100-ft (30-m).
$1,516.86 - $3,841.86CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Only RK-59353-70 Mfr # 626650
COMPARE
Handheld Optical Dissolved Oxygen Meter Only
$1,516.86CAD / Each
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Kit, ODO/T Probe with 4-m Cable RK-59353-85 Mfr # 626663
COMPARE
Handheld Optical Dissolved Oxygen Meter Kit, ODO/T Probe with 4-m Cable
$2,833.33CAD / Each
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Kit, ODO/T Probe with 10-m Cable RK-59353-86 Mfr # 626664
COMPARE
Handheld Optical Dissolved Oxygen Meter Kit, ODO/T Probe with 10-m Cable
$2,925.42CAD / Each
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Kit, ODO/CT Probe with 4-m Cable RK-59353-87 Mfr # 626660
COMPARE
Handheld Optical Dissolved Oxygen Meter Kit, ODO/CT Probe with 4-m Cable
$3,733.94CAD / Each
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Kit, ODO/CT Probe with 10-m Cable RK-59353-88 Mfr # 626661
COMPARE
Handheld Optical Dissolved Oxygen Meter Kit, ODO/CT Probe with 10-m Cable
$3,841.86CAD / Each