Sherwood Scientific 50186700 Software for Fluid Bed Dryers
Representative image only.
Sherwood Scientific 50186700 Software for Fluid Bed Dryers
Sherwood Scientific | Mfr: 50186700| Item: 0265822
Optimize performance of your Sherwood Scientific fluid bed dryer
$1,510.22CAD / Each
$1,510.22CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50186700 Software for Fluid Bed Dryers
Sherwood Scientific 50186700 Software for Fluid Bed Dryers
0265822
$1,510.22CAD / Each
+ + + + + + =
Total Price 40032.37 ADD 7 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your fluid bed dryer running properly with these accessories
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Sherwood Scientific fluid bed dryer.
Software allows direct control of the dryer. Write a program with any number of steps, controlling the time and inlet temperature of each step. End each program step after a time interval, at a temperature point, or at a humidity level. Requires IBM® compatible computer running Windows® 98 or higher, with a Pentium processor and 32 MB of RAM.
Specifications
  • DescriptionSoftware for Fluid Bed Dryers