Sherwood Scientific 50035407 Filter Bag for Non-Sealed Tubs, Large, Nomex
Representative image only.
Sherwood Scientific 50035407 Filter Bag for Non-Sealed Tubs, Large, Nomex
Sherwood Scientific | Mfr: 50035407| Item: 0265881
Safely retains stray particles
$504.64CAD / Each
$504.64CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50035407 Filter bag for non-sealed tubs, Large, Nomex
Sherwood Scientific 50035407 Filter bag for non-sealed tubs, Large, Nomex
0265881
$504.64CAD / Each
+ + + =
Total Price 11127.78 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Allows passage of exit gases
More About this Item
Bags retain stray particles of the sample being fluidized in the tub while allowing the passage of exit gases. Use one large bag for a single two- or five-liter tub assembly; use four small bags for the multi-tub assemblies.
Specifications
  • DescriptionFilter bag for non-sealed tubs, Large, Nomex