Sherwood Scientific 50135020 Sealed Tub Assembly, with port for temperature/humidity probe; 5-liter
Representative image only.
Sherwood Scientific 50135020 Sealed Tub Assembly, with port for temperature/humidity probe; 5-liter
Sherwood Scientific | Mfr: 50135020| Item: 0265801
Reduce sample loss
$7,867.90CAD / Each
$7,867.90CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50135020 Sealed Tub Assembly, with port for temperature/humidity probe; 5-liter
Sherwood Scientific 50135020 Sealed Tub Assembly, with port for temperature/humidity probe; 5-liter
0265801
$7,867.90CAD / Each
+ + + =
Total Price 38426.01 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Silicon O-ring provides tight seal
  • 1 year warranty
More About this Item
Sealed tubs reduce sample loss - ideal for drying smaller particles. To avoid overflow of sample into the filter bag, the filter plate seals on a silicon O-ring, retaining the sample in the tub.
Includes a port to add extra sample and a port for a temperature/humidty probe. Optional filter top caps are sold separately.
1 Year Warranty
Specifications
  • Materials of ConstructionGlass
  • DescriptionSealed Tub Assembly, with port for temperature/humidity probe; 5-liter