Sherwood Scientific 50035021 Filter top cap for 5 L sealed tubs, 250 mesh

Representative image only.

Sherwood Scientific 50035021 Filter top cap for 5 L sealed tubs, 250 mesh

Sherwood Scientific | Mfr: 50035021| Item: 0265866

$1,554.43CAD / Each

$1,554.43CAD / Each

Buy It Together

Current Item

Sherwood Scientific 50035021 Filter top cap for 5 L sealed tubs, 250 mesh
Sherwood Scientific 50035021 Filter top cap for 5 L sealed tubs, 250 mesh
0265866

$1,554.43CAD / Each
+ + + + =
Total Price 35306.11 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Select cap 02658-68 for wetter or denser samples requiring more air flow. Use filter top cap 02658-69 for drying 5-µm particles.

Note: Only 100 g of sample can be dried at a time with filter top cap 02658-69.

Specifications
  • DescriptionFilter top cap for 5 L sealed tubs, 250 mesh
Reviews
Q&A