Sherwood Scientific 50186001 Air Pulse Flow Module for Fluid Bed Dryer
Representative image only.
Sherwood Scientific 50186001 Air Pulse Flow Module for Fluid Bed Dryer
Sherwood Scientific | Mfr: 50186001| Item: 0265821
Optimize performance of your Sherwood Scientific fluid bed dryer
$2,622.63CAD / Each
$2,622.63CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50186001 Air Pulse Flow Module for Fluid Bed Dryer
Sherwood Scientific 50186001 Air Pulse Flow Module for Fluid Bed Dryer
0265821
$2,622.63CAD / Each
+ + + + + =
Total Price 40971.29 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your fluid bed dryer running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Sherwood Scientific fluid bed dryer.
Pulser unit fits on the back of the unit and pulses the air flow to break up clumps of moist material. Pulse frequency can be varied.
1 Year Warranty
Specifications
  • DescriptionAir Pulse Flow Module for Fluid Bed Dryer