Sherwood Scientific 50035023 Filter top cap for 5-L sealed tub with nylon mesh filter bag
Representative image only.
Sherwood Scientific 50035023 Filter top cap for 5-L sealed tub with nylon mesh filter bag
Sherwood Scientific | Mfr: 50035023| Item: 0265868
$1,280.38CAD / Each
$1,280.38CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50035023 Filter top cap for 5-L sealed tub with nylon mesh filter bag
Sherwood Scientific 50035023 Filter top cap for 5-L sealed tub with nylon mesh filter bag
0265868
$1,280.38CAD / Each
+ + + + =
Total Price 35032.06 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Select cap 02658-68 for wetter or denser samples requiring more air flow. Use filter top cap 02658-69 for drying 5-µm particles.
Note: Only 100 g of sample can be dried at a time with filter top cap 02658-69.
Specifications
  • DescriptionFilter top cap for 5-L sealed tub with nylon mesh filter bag