Sherwood Scientific 50035022 Filter top cap for 5-L sealed tub, 500 mesh
Representative image only.
Sherwood Scientific 50035022 Filter top cap for 5-L sealed tub, 500 mesh
Sherwood Scientific | Mfr: 50035022| Item: 0265867
$1,885.94CAD / Each
$1,885.94CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50035022 Filter top cap for 5-L sealed tub, 500 mesh
Sherwood Scientific 50035022 Filter top cap for 5-L sealed tub, 500 mesh
0265867
$1,885.94CAD / Each
+ + + + =
Total Price 35637.62 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Select cap 02658-68 for wetter or denser samples requiring more air flow. Use filter top cap 02658-69 for drying 5-µm particles.
Note: Only 100 g of sample can be dried at a time with filter top cap 02658-69.
Specifications
  • DescriptionFilter top cap for 5-L sealed tub, 500 mesh