Sherwood Scientific 50035014 Sealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass
Representative image only.
Sherwood Scientific 50035014 Sealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass
Sherwood Scientific | Mfr: 50035014| Item: 0265864
Reduce sample loss
$5,996.70CAD / Each
$5,996.70CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50035014 Sealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass
Sherwood Scientific 50035014 Sealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass
0265864
$5,996.70CAD / Each
+ + + =
Total Price 36554.81 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Silicon O-ring provides tight seal
More About this Item
Sealed tubs reduce sample loss - ideal for drying smaller particles. To avoid overflow of sample into the filter bag, the filter plate seals on a silicon O-ring, retaining the sample in the tub.
Specifications
  • Materials of ConstructionGlass
  • DescriptionSealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass