Sherwood Scientific 50035009 5-Liter, Glass Tub and Assembly for Fluid Bed Dryer
Representative image only.
Sherwood Scientific 50035009 5-Liter, Glass Tub and Assembly for Fluid Bed Dryer
Sherwood Scientific | Mfr: 50035009| Item: 0265856
Quickly and evenly dry particles as small as 5 μm
$3,838.18CAD / Each
$3,838.18CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50035009 5-Liter, Glass Tub and Assembly for Fluid Bed Dryer
Sherwood Scientific 50035009 5-Liter, Glass Tub and Assembly for Fluid Bed Dryer
0265856
$3,838.18CAD / Each
+ + + + + =
Total Price 35251.23 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials
  • Variety of accessories provide easy customization for your drying applications
More About this Item
Non-Sealed Tub Assemblies are available in glass or stainless steel and require a filter bag, sold separately below.
Model 02658-02 includes a port to add extra sample and a port for a temperature/humidity probe (sold separately).
Order fluid bed dryer, filter bags, and optional accessories separately for a complete fluid bed dryer system.
Glass tubs are useful for observation of applications and determination of the optimum fluidizing velocity. The 2- and 5-L units handle samples up to 11 lb (5.0 kg). Required filter bags sold separately.
Specifications
  • Materials of ConstructionGlass
  • Description5-Liter, Glass Tub and Assembly for Fluid Bed Dryer