Sherwood Scientific 50035400 Filter bag for non-sealed tubs, Large, Nylon
Representative image only.
Sherwood Scientific 50035400 Filter bag for non-sealed tubs, Large, Nylon
Sherwood Scientific | Mfr: 50035400| Item: 0265882
Safely retains stray particles
$350.30CAD / Each
$350.30CAD / Each
Buy It Together
Current Item
Sherwood Scientific 50035400 Filter bag for non-sealed tubs, Large, Nylon
Sherwood Scientific 50035400 Filter bag for non-sealed tubs, Large, Nylon
0265882
$350.30CAD / Each
+ + + =
Total Price 10973.44 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Allows passage of exit gases
More About this Item
Bags retain stray particles of the sample being fluidized in the tub while allowing the passage of exit gases. Use one large bag for a single two- or five-liter tub assembly; use four small bags for the multi-tub assemblies.
Specifications
  • DescriptionFilter bag for non-sealed tubs, Large, Nylon