Sherwood Scientific 500 350 49 Low Density Classifier Assembly for Fluid Bed Dryer
Representative image only.
Sherwood Scientific 500 350 49 Low Density Classifier Assembly for Fluid Bed Dryer
Sherwood Scientific | Mfr: 50035049| Item: 0265876
Optimize performance of your Sherwood Scientific fluid bed dryer
$14,439.22CAD / Each
$14,439.22CAD / Each
Buy It Together
Current Item
Sherwood Scientific 500 350 49 Low Density Classifier Assembly for Fluid Bed Dryer
Sherwood Scientific 500 350 49 Low Density Classifier Assembly for Fluid Bed Dryer
0265876
$14,439.22CAD / Each
+ + + + + =
Total Price 52787.88 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your fluid bed dryer running properly with these accessories
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Sherwood Scientific fluid bed dryer.
Low-Density Classifier Assembly lets you take a representative sample in seconds without interrupting the drying process. Separates particles of your sample by size, shape, and moisture density. Selectively remove particles from the sample by adjusting the air flow rate - the first particles to dry will separate first since they are less dense. Use the low-density assembly for light particles like organic powders and pharmaceuticals.
Specifications
  • DescriptionLow Density Classifier Assembly for Fluid Bed Dryer