Hach TL2300 Tungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU
Representative image only.
Hach TL2300 Tungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU
Hach | Mfr: LPV444.99.00210| Item: 9951225
Simplify testing in demanding industrial and wastewater applications
$6,295.80CAD / Each
$6,295.80CAD / Each
Buy It Together
Current Item
Hach TL2300 Tungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU
Hach TL2300 Tungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU
9951225
$6,295.80CAD / Each
+ + =
Total Price 7303.60 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
USEPA filter assembly, six 1-inch sample cells (30 mL) with caps, Gelex® secondary turbidity standardization kit, StablCal® calibration kit, silicone oil, oiling cloth, power supply, power cord, and dust cover.
Key Features
  • Identifies stable readings to ensure accurate analysis
  • Displays measurements more clearly on large, color touch screen
  • Locks in measurements and logs up to 2000 data points
  • Easily exports data via a USB port
  • EPA Compliant
  • 1 year warranty
More About this Item
The TL23 series benchtop turbidimeters simplify the measurement of high range turbidity applications with an easy-to-use color touch screen and intuitive user interface. Features include self-diagnostics for quality assurance, single-point calibration for measurements under 40 NTU/FNU, and sample identification for traceability.
Stable readings and accurate analysis are ensured by capturing turbidity reading once the turbidimeter detects sample stability. No need for repeat measurements.
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Meter TypeBenchtop
  • Display typeColor touch screen
  • Min Turbidity0
  • Max Turbidity4000
  • Turbidity Resolution0.001 NTU/EBC
  • Turbidity Accuracy±2% of reading plus 0.01 NTU from 0 to 1000 NTU, ±5% of reading from 1000 to 4000 NTU
  • OutputUSB
  • Data Logging (points)2000
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionTungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU