Hach Stablcal® Turbidity Standards Calibration Kit, 100 mL Bottles
Representative image only.
Hach Stablcal® Turbidity Standards Calibration Kit, 100 mL Bottles
Hach | Mfr: 2662110| Item: 9951221
Optimize water analysis
$546.63CAD / Each
$546.63CAD / Each
Buy It Together
Current Item
Hach Stablcal® Turbidity Standards Calibration Kit, 100 mL Bottles
Hach Stablcal® Turbidity Standards Calibration Kit, 100 mL Bottles
9951221
$546.63CAD / Each
+
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
+ +
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU
9951220
$6,375.36CAD / Each
+
Hach 2100Q  Handheld Turbidity Meter, EPA Compliant
9951110
$3,166.32CAD / Each
+ =
Total Price 11063.99 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Kit includes 100 mL bottles of <0.1, 20, 200, 1000 and 4000 NTU standards.
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
1 Year Warranty
Specifications
  • Compatible WithHach Turbidity meters
  • DescriptionStablcal turbidity standards calibration kit for TL23 series, 100 mL bottles